siRNA Details
Gene Symbol: NOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267721.1 | NP_001254650.1 | 3 | 576 | ||
NM_144603.3 | NP_653204.1 | 3 | 576 | |||
NM_172167.2 | NP_751907.1 | 3 | 579 | |||
NM_172168.2 | NP_751908.1 | 3 | 579 | |||
XM_005255099.4 | XP_005255156.1 | 4 | 1674 | |||
XM_017022927.1 | XP_016878416.1 | 4 | 1180 | |||
XM_017022928.1 | XP_016878417.1 | 4 | 537 | |||
GenBank mRNA | AB097667.1 | - | 3 | 389 | ||
AF532983.1 | - | 3 | 210 | |||
AF532984.1 | - | 3 | 213 | |||
AF532985.1 | - | 3 | 213 | |||
AF539796.1 | - | 3 | 209 | |||
AK292197.1 | - | 3 | 579 | |||
AK310198.1 | - | 2 | 670 | |||
AK313917.1 | - | 3 | 499 | |||
AY191359.1 | - | 3 | 210 | |||
AY255768.1 | - | 3 | 206 | |||
BC015917.2 | - | 3 | 466 | 4661469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
