siRNA Details
Gene Symbol: FGF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003868.2 | NP_003859.1 | 3 | 491 | ||
GenBank mRNA | AB009391.1 | - | 3 | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
