siRNA Details
Gene Symbol: SLC26A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167962.1 | NP_001161434.1 | 6 | 689 | ||
NM_001321787.1 | NP_001308716.1 | 6 | 689 | |||
NM_198999.2 | NP_945350.1 | 6 | 689 | |||
NM_206883.2 | NP_996766.1 | 6 | 689 | |||
NM_206884.2 | NP_996767.1 | 6 | 689 | |||
NM_206885.2 | NP_996768.1 | 6 | 689 | |||
NR_120441.1 | 5 | 609 | ||||
NR_120442.1 | 5 | 609 | ||||
NR_120443.1 | 5 | 609 | ||||
NR_135801.1 | 6 | 689 | ||||
NR_135802.1 | 6 | 689 | ||||
XM_011516170.2 | XP_011514472.1 | 6 | 1479 | |||
GenBank mRNA | AF523354.1 | - | 6 | 663 | ||
AK126898.1 | - | 5 | 578 | |||
AY256823.1 | - | 6 | 663 | |||
AY256824.1 | - | 6 | 663 | |||
AY256825.1 | - | 6 | 663 | |||
AY289133.1 | - | 4 | 435 | |||
AY289134.1 | - | 4 | 435 | |||
BC100832.1 | - | 5 | 594 | 40001465 | ||
BC100833.1 | - | 5 | 594 | 40001467 | ||
BC100834.1 | - | 5 | 594 | |||
BC100835.1 | - | 5 | 594 | 40001470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
