siRNA Details
Gene Symbol: NLRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033053.2 | NP_001028225.1 | 11, 12 | 3869 | ||
NM_014922.4 | NP_055737.1 | 11, 12 | 3857 | |||
NM_033004.3 | NP_127497.1 | 11, 12 | 3857 | |||
NM_033006.3 | NP_127499.1 | 10, 11 | 3767 | |||
NM_033007.3 | NP_127500.1 | 10, 11 | 3767 | |||
GenBank mRNA | AB023143.1 | - | 11, 12 | 3824 | ||
AF229059.1 | - | 11, 12 | 3302 | |||
AF229060.1 | - | 11, 12 | 3302 | |||
AF229061.1 | - | 10, 11 | 3212 | |||
AF229062.1 | - | 10, 11 | 3212 | |||
AF298548.1 | - | 12, 13 | 3461 | |||
AF310105.1 | - | 11, 12 | 3820 | |||
AK026393.1 | - | 8, 9 | 1780 | |||
AK026398.1 | - | 8, 9 | 1130 | |||
AK310936.1 | - | 12, 13 | 3668 | |||
AL117470.1 | - | 8, 9 | 2460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
