siRNA Details
Gene Symbol: NLRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033053.2 | NP_001028225.1 | 15 | 4541 | ||
NM_014922.4 | NP_055737.1 | 14 | 4397 | |||
NM_033004.3 | NP_127497.1 | 15 | 4529 | |||
NM_033006.3 | NP_127499.1 | 14 | 4439 | |||
NM_033007.3 | NP_127500.1 | 13 | 4307 | |||
GenBank mRNA | AB023143.1 | - | 14 | 4364 | ||
AF229059.1 | - | 14 | 3842 | |||
AF229060.1 | - | 15 | 3974 | |||
AF229061.1 | - | 13 | 3752 | |||
AF229062.1 | - | 14 | 3884 | |||
AF298548.1 | - | 15 | 4001 | |||
AF310105.1 | - | 14 | 4360 | |||
AK026393.1 | - | 12 | 2453 | |||
AK026398.1 | - | 12 | 1802 | |||
AL117470.1 | - | 12 | 3132 | |||
AL832400.1 | - | 1 | 738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
