siRNA Details
Gene Symbol: NLRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033053.2 | NP_001028225.1 | 9 | 3574 | ||
NM_014922.4 | NP_055737.1 | 9 | 3574 | |||
NM_033004.3 | NP_127497.1 | 9 | 3574 | |||
NM_033006.3 | NP_127499.1 | 8 | 3484 | |||
NM_033007.3 | NP_127500.1 | 8 | 3484 | |||
GenBank mRNA | AB023143.1 | - | 9 | 3541 | ||
AF229059.1 | - | 9 | 3019 | |||
AF229060.1 | - | 9 | 3019 | |||
AF229061.1 | - | 8 | 2929 | |||
AF229062.1 | - | 8 | 2929 | |||
AF298548.1 | - | 10 | 3178 | |||
AF310105.1 | - | 9 | 3537 | |||
AK026393.1 | - | 6 | 1497 | |||
AK026398.1 | - | 6 | 835 | |||
AK310936.1 | - | 10 | 3385 | |||
AL117470.1 | - | 6 | 2177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
