siRNA Details
Gene Symbol: LTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000627.3 | NP_000618.3 | 12 | 1827 | ||
NM_001166264.1 | NP_001159736.1 | 12 | 1827 | |||
NM_001166265.1 | NP_001159737.1 | 12 | 1668 | |||
NM_001166266.1 | NP_001159738.1 | 12 | 1668 | |||
NM_206943.2 | NP_996826.2 | 16 | 2687 | |||
XM_005264317.3 | XP_005264374.1 | 16 | 2530 | |||
XM_005264318.3 | XP_005264375.1 | 16 | 2530 | |||
XM_011532853.2 | XP_011531155.1 | 16 | 2863 | |||
XM_011532855.2 | XP_011531157.1 | 16 | 2686 | |||
XM_011532856.2 | XP_011531158.1 | 16 | 2692 | |||
XM_011532857.2 | XP_011531159.1 | 16 | 2692 | |||
XM_011532859.2 | XP_011531161.1 | 16 | 2533 | |||
XM_011532860.2 | XP_011531162.1 | 15 | 2524 | |||
XM_011532861.2 | XP_011531163.1 | 16 | 2692 | |||
XM_011532862.2 | XP_011531164.1 | 16 | 2692 | |||
XM_017004108.1 | XP_016859597.1 | 16 | 2689 | |||
XM_017004109.1 | XP_016859598.1 | 16 | 2689 | |||
XM_017004110.1 | XP_016859599.1 | 16 | 2689 | |||
GenBank mRNA | AB208801.1 | - | 12 | 1626 | ||
AF489528.1 | - | 12 | 1683 | |||
AK298027.1 | - | 6 | 396 | |||
AK299912.1 | - | 7 | 625 | |||
AL703596.1 | - | 6 | 518 | |||
BC130289.1 | - | 12 | 1767 | 40146320 | ||
BC144127.1 | - | 12 | 1610 | |||
BC144128.1 | - | 12 | 1767 | 9052648 | ||
M34057.1 | - | 12 | 1773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
