siRNA Details
Gene Symbol: ATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127713.1 | NP_001121185.1 | 13 | 1874 | ||
NM_015915.4 | NP_056999.2 | 12 | 1690 | |||
NM_181598.3 | NP_853629.2 | 12 | 1690 | |||
GenBank mRNA | AF131801.1 | - | 11 | 1214 | ||
AF444143.1 | - | 12 | 1621 | |||
AK223436.1 | - | 12 | 1603 | |||
AK290185.1 | - | 12 | 1603 | |||
AK312518.1 | - | 12 | 1576 | |||
AL833591.1 | - | 13 | 1658 | |||
AY032844.1 | - | 12 | 1618 | |||
BC010708.2 | - | 12 | 1593 | 3869877 | ||
CR457153.1 | - | 11 | 1200 | |||
KU178621.1 | - | 12 | 1449 | |||
KU178622.1 | - | 12 | 1449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
