siRNA Details
Gene Symbol: OR8B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005468.1 | NP_001005468.1 | 1 | 694 | ||
XM_017017535.1 | XP_016873024.1 | 3 | 926 | |||
XM_017017536.1 | XP_016873025.1 | 3 | 912 |
Gene Symbol: OR8B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005467.1 | NP_001005467.1 | 1 | 694 | ||
XM_017017716.1 | XP_016873205.1 | 6 | 2060 | |||
XM_017017717.1 | XP_016873206.1 | 6 | 1363 | |||
XM_017017718.1 | XP_016873207.1 | 4 | 1791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
