siRNA Details
Gene Symbol: ZDHHC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001483.2 | NP_001001483.1 | 14 | 1479 | ||
NM_019028.2 | NP_061901.2 | 15 | 1625 | |||
XM_005252996.2 | XP_005253053.1 | 15 | 1597 | |||
XM_011520193.1 | XP_011518495.1 | 12 | 1238 | |||
XM_011520194.2 | XP_011518496.1 | 16 | 1744 | |||
XM_011520195.1 | XP_011518497.1 | 15 | 1535 | |||
GenBank mRNA | AB024495.1 | - | 15 | 1573 | ||
AB097014.1 | - | 15 | 1623 | |||
AB097027.1 | - | 15 | 1623 | |||
AK001714.1 | - | 15 | 1623 | |||
AK001831.1 | - | 10 | 997 | |||
AK123539.1 | - | 1 | 1326 | |||
AK291348.1 | - | 15 | 1600 | |||
AL833714.1 | - | 11 | 1075 | |||
BC036020.1 | - | 15 | 1610 | 5272445 | ||
BC050690.1 | - | 14 | 1479 | 6171322 | ||
BC056152.1 | - | 15 | 1625 | 6170701 | ||
BX648156.1 | - | 13 | 5491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
