siRNA Details
Gene Symbol: ZDHHC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001483.2 | NP_001001483.1 | 8 | 893 | ||
NM_019028.2 | NP_061901.2 | 9 | 1039 | |||
XM_005252996.2 | XP_005253053.1 | 9 | 1011 | |||
XM_011520193.1 | XP_011518495.1 | 7 | 736 | |||
XM_011520194.2 | XP_011518496.1 | 9 | 1014 | |||
XM_011520195.1 | XP_011518497.1 | 9 | 949 | |||
GenBank mRNA | AB024495.1 | - | 9 | 987 | ||
AB097014.1 | - | 9 | 1037 | |||
AB097027.1 | - | 9 | 1037 | |||
AK001714.1 | - | 9 | 1037 | |||
AK001831.1 | - | 4 | 411 | |||
AK291348.1 | - | 9 | 1014 | |||
AL833714.1 | - | 5 | 489 | |||
BC036020.1 | - | 9 | 1024 | 5272445 | ||
BC050690.1 | - | 8 | 893 | 6171322 | ||
BC056152.1 | - | 9 | 1039 | 6170701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
