siRNA Details
Gene Symbol: HES5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010926.3 | NP_001010926.1 | 1 | 100 | ||
XM_005244751.4 | XP_005244808.1 | 1 | 409 | |||
GenBank mRNA | BC087840.1 | - | 1 | 100 | 6204648 | |
DQ272660.1 | - | 1 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
