siRNA Details
Gene Symbol: NFATC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136021.2 | NP_001129493.1 | 5 | 1703 | ||
NM_001258292.1 | NP_001245221.1 | 5 | 1703 | |||
NM_001258294.1 | NP_001245223.1 | 5 | 1189 | |||
NM_001258295.1 | NP_001245224.1 | 5 | 1189 | |||
NM_001258296.1 | NP_001245225.1 | 5 | 1267 | |||
NM_001258297.1 | NP_001245226.1 | 5 | 1267 | |||
NM_012340.4 | NP_036472.2 | 5 | 1781 | |||
NM_173091.3 | NP_775114.1 | 5 | 1781 | |||
XM_011528824.2 | XP_011527126.1 | 5 | 1772 | |||
XM_011528825.2 | XP_011527127.1 | 5 | 1729 | |||
XM_011528826.2 | XP_011527128.1 | 5 | 1210 | |||
XM_017027850.1 | XP_016883339.1 | 5 | 1005 | |||
XM_017027851.1 | XP_016883340.1 | 5 | 1772 | |||
GenBank mRNA | AK226146.1 | - | 5 | 1703 | ||
BC136418.1 | - | 5 | 1684 | 9020406 | ||
BC144074.1 | - | 5 | 1684 | 9052594 | ||
BM463165.1 | - | 572 | ||||
EU887573.1 | - | 5 | 1561 | |||
EU887574.1 | - | 5 | 1501 | |||
EU887575.1 | - | 5 | 1189 | |||
EU887576.1 | - | 5 | 1267 | |||
EU887577.1 | - | 5 | 1501 | |||
EU887578.1 | - | 5 | 1561 | |||
EU887579.1 | - | 5 | 1189 | |||
EU887580.1 | - | 5 | 1267 | |||
U43341.1 | - | 5 | 1781 | |||
U43342.1 | - | 5 | 1781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
