siRNA Details
Gene Symbol: LY6G6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003673.3 | 1, 2 | 310 | |||
NR_024541.1 | 1, 2 | 310 | ||||
GenBank mRNA | AJ245419.1 | - | 1, 2 | 57 | ||
AJ315538.1 | - | 1, 2 | 57 | |||
AJ315539.1 | - | 1, 2 | 57 | |||
BC069141.1 | - | 1, 2 | 57 | |||
DA786728.1 | - | 1, 2 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
