siRNA Details
Gene Symbol: LY6G6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003673.3 | 3 | 499 | |||
GenBank mRNA | AJ245419.1 | - | 3 | 225 | ||
AJ315538.1 | - | 3 | 246 | |||
AJ315539.1 | - | 2 | 348 | |||
BC069141.1 | - | 3 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
