siRNA Details
Gene Symbol: COPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012133.5 | NP_036265.3 | 22 | 2430 | ||
GenBank mRNA | AB047847.1 | - | 22 | 2340 | ||
AF157833.1 | - | 22 | 2400 | |||
AF207598.1 | - | 22 | 2396 | |||
AF308295.1 | - | 3 | 293 | |||
AK002086.1 | - | 14 | 1732 | |||
AK022934.1 | - | 22 | 2327 | |||
AK291409.1 | - | 22 | 2420 | |||
BC017443.1 | - | 22 | 2323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
