siRNA Details
Gene Symbol: CCDC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008391.3 | NP_001008392.2 | 15 | 1378 | ||
XM_011520139.2 | XP_011518441.1 | 14 | 1239 | |||
GenBank mRNA | AK128159.1 | - | 6 | 597 | ||
AK131470.1 | - | 15 | 1361 | |||
BC062749.1 | - | 15 | 1348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
