siRNA Details
Gene Symbol: RAPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203365.3 | NP_976241.1 | 4 | 535 | ||
NM_213589.2 | NP_998754.1 | 4 | 535 | |||
XM_005246777.3 | XP_005246834.1 | 4 | 530 | |||
XM_005246779.4 | XP_005246836.1 | 4 | 1429 | |||
XM_006712695.3 | XP_006712758.1 | 5 | 1510 | |||
XM_011511646.2 | XP_011509948.1 | 5 | 602 | |||
XM_011511647.1 | XP_011509949.1 | 3 | 310 | |||
XM_011511648.1 | XP_011509950.1 | 3 | 310 | |||
XM_011511649.2 | XP_011509951.1 | 4 | 530 | |||
GenBank mRNA | AB051468.1 | - | 3 | 268 | ||
AB053311.1 | - | 4 | 529 | |||
AJ584699.1 | - | 4 | 529 | |||
AK096270.1 | - | 4 | 479 | |||
AY494951.1 | - | 4 | 531 | |||
AY523977.1 | - | 4 | 531 | |||
AY523978.1 | - | 4 | 531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
