siRNA Details
Gene Symbol: ARHGAP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004491.4 | NP_004482.4 | 6 | 8284 | ||
XM_017026714.1 | XP_016882203.1 | 7 | 8680 | |||
GenBank mRNA | AK097314.1 | - | 1 | 1182 | ||
BC003514.1 | - | 1 | 2683 | |||
BC065491.1 | - | 1 | 1672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
