siRNA Details
Gene Symbol: TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003823.3 | NP_003814.1 | 1 | 207 | ||
GenBank mRNA | AF104419.1 | - | 1 | 190 | ||
AF134240.1 | - | 1 | 90 | |||
AF217794.1 | - | 1 | 142 | |||
AY124378.1 | - | 1 | 129 | |||
AY124379.1 | - | 1 | 129 | |||
AY358279.1 | - | 1 | 190 | |||
BC017065.1 | - | 1 | 189 | 3886635 | ||
BC034349.1 | - | 1 | 198 | 4752507 | ||
BM759033.1 | - | 1 | 209 |
Gene Symbol: RTEL1-TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037882.1 | 36 | 4824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
