siRNA Details
Gene Symbol: TSPAN17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006616.2 | NP_001006617.2 | 1 | 290 | ||
NM_012171.2 | NP_036303.1 | 1 | 290 | |||
NM_130465.4 | NP_569732.2 | 1 | 290 | |||
XM_005265876.3 | XP_005265933.1 | 1 | 270 | |||
GenBank mRNA | AF174603.1 | - | 1 | 97 | ||
AK024427.1 | - | 1 | 180 | |||
AY037146.1 | - | 1 | 210 | |||
BC010405.2 | - | 1 | 112 | 3621871 | ||
BC067105.1 | - | 1 | 175 | 6646576 | ||
BI756001.1 | - | 1 | 252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
