siRNA Details
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 7 | 927 | ||
XM_011523496.2 | XP_011521798.1 | 8 | 983 | |||
XM_011523498.2 | XP_011521800.1 | 8 | 1017 | |||
XM_011523501.2 | XP_011521803.1 | 4 | 809 | |||
XM_017023248.1 | XP_016878737.1 | 8 | 1420 | |||
XM_017023249.1 | XP_016878738.1 | 7 | 1237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
