siRNA Details
Gene Symbol: SHISA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145176.1 | NP_001138648.1 | 4 | 3663 | ||
GenBank mRNA | AK094959.1 | - | 1 | 282 | ||
AK127223.1 | - | 1 | 1834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
