siRNA Details
Gene Symbol: PIGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012327.5 | NP_036459.1 | 22 | 2477 | ||
NM_176787.4 | NP_789744.1 | 23 | 2554 | |||
XM_011525889.1 | XP_011524191.1 | 23 | 2522 | |||
XM_011525890.1 | XP_011524192.1 | 23 | 2719 | |||
XM_011525891.1 | XP_011524193.1 | 23 | 2437 | |||
XM_011525892.1 | XP_011524194.1 | 22 | 2459 | |||
XM_011525893.1 | XP_011524195.1 | 22 | 2410 | |||
XM_011525894.1 | XP_011524196.1 | 22 | 2321 | |||
XM_011525895.1 | XP_011524197.1 | 21 | 2319 | |||
XM_011525896.1 | XP_011524198.1 | 23 | 2522 | |||
XM_011525898.1 | XP_011524200.1 | 23 | 2522 | |||
XM_017025685.1 | XP_016881174.1 | 20 | 2120 | |||
XM_017025686.1 | XP_016881175.1 | 17 | 1710 | |||
GenBank mRNA | AF109219.1 | - | 22 | 2390 | ||
AK000730.1 | - | 8 | 968 | |||
AK315134.1 | - | 23 | 2522 | |||
AL137607.1 | - | 19 | 1789 | |||
BC028363.2 | - | 23 | 2491 | 4826763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
