siRNA Details
Gene Symbol: INPP5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017915.2 | NP_001017915.1 | 2 | 681 | ||
NM_005541.4 | NP_005532.2 | 2 | 681 | |||
XM_017004004.1 | XP_016859493.1 | 2 | 304 | |||
GenBank mRNA | AK298651.1 | - | 2 | 310 | ||
AK309644.1 | - | 2 | 306 | |||
BC099920.1 | - | 2 | 292 | 6304992 | ||
BC113580.1 | - | 2 | 229 | 8322632 | ||
BC113582.1 | - | 2 | 229 | 8322634 | ||
U50041.1 | - | 2 | 290 | |||
U53470.1 | - | 2 | 299 | |||
U57650.1 | - | 2 | 681 | |||
U84400.1 | - | 2 | 250 | |||
X98429.1 | - | 2 | 309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
