siRNA Details
Gene Symbol: ZNF536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014717.2 | NP_055532.1 | 5 | 4656 | ||
XM_017027539.1 | XP_016883028.1 | 8 | 5268 | |||
XM_017027542.1 | XP_016883031.1 | 7 | 5180 | |||
GenBank mRNA | AB002388.1 | - | 5 | 4628 | ||
AK091070.1 | - | 2133 | ||||
AK128073.1 | - | 1 | 3795 | |||
BC146757.1 | - | 5 | 4628 | 9007195 | ||
BC150171.1 | - | 5 | 4635 | 40147081 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
