siRNA Details
Gene Symbol: SLC35E2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110781.2 | NP_001104251.1 | 6 | 1225 | ||
NM_001290264.1 | NP_001277193.1 | 7 | 1643 | |||
GenBank mRNA | AB007916.3 | - | 6 | 1163 | ||
AK125619.1 | - | 6 | 1149 | |||
BC110653.1 | - | 6 | 1201 | 4392812 | ||
CR936618.1 | - | 7 | 1363 | |||
DA794179.1 | - | 3 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
