siRNA Details
Gene Symbol: ANKFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257999.1 | NP_001244928.1 | 7 | 1047 | ||
NM_016376.3 | NP_057460.3 | 7 | 842 | |||
NR_047571.1 | 6 | 723 | ||||
XM_005256679.3 | XP_005256736.1 | 7 | 889 | |||
XM_011523926.1 | XP_011522228.1 | 7 | 945 | |||
XM_011523928.2 | XP_011522230.1 | 7 | 927 | |||
XM_011523933.1 | XP_011522235.1 | 3 | 331 | |||
XM_017024733.1 | XP_016880222.1 | 7 | 1142 | |||
GenBank mRNA | AB033081.2 | - | 7 | 1047 | ||
AK025483.1 | - | 7 | 819 | |||
AK057047.1 | - | 6 | 700 | |||
AK225170.1 | - | 7 | 819 | |||
BC052308.1 | - | 7 | 815 | |||
CR933717.1 | - | 1 | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
