siRNA Details
Gene Symbol: KLHL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020805.2 | NP_065856.1 | 2 | 1233 | ||
XM_011526109.2 | XP_011524411.2 | 2 | 925 | |||
GenBank mRNA | AB037805.1 | - | 2 | 1233 | ||
AK294996.1 | - | 1 | 471 | |||
BC021267.2 | - | 2 | 1101 | 4588702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
