siRNA Details
Gene Symbol: KLHL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020805.2 | NP_065856.1 | 6 | 1795 | ||
GenBank mRNA | AB037805.1 | - | 6 | 1795 | ||
AK294996.1 | - | 5 | 1033 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
