siRNA Details
Gene Symbol: RPL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000984.5 | NP_000975.2 | 5 | 699 | ||
GenBank mRNA | BC058041.1 | - | 5 | 695 | 6716166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
