siRNA Details
Gene Symbol: PER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022817.2 | NP_073728.1 | 11 | 1461 | ||
XM_005246111.4 | XP_005246168.1 | 11 | 1447 | |||
XM_006712824.3 | XP_006712887.1 | 11 | 1399 | |||
GenBank mRNA | AB002345.2 | - | 11 | 1346 | ||
AB012614.1 | - | 9 | 1117 | |||
AK294876.1 | - | 11 | 1461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
