siRNA Details
Gene Symbol: SH3PXD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014631.2 | NP_055446.2 | 14 | 10991 | ||
XM_005270294.4 | XP_005270351.1 | 15 | 11124 | |||
XM_005270295.4 | XP_005270352.1 | 14 | 11078 | |||
XM_005270297.3 | XP_005270354.1 | 9 | 12355 | |||
XM_005270298.4 | XP_005270355.1 | 9 | 11158 | |||
XM_006718079.3 | XP_006718142.1 | 16 | 11221 | |||
XM_006718080.3 | XP_006718143.1 | 10 | 11252 | |||
XM_011540385.2 | XP_011538687.1 | 15 | 11175 | |||
XM_011540386.2 | XP_011538688.1 | 10 | 12451 | |||
XM_011540387.2 | XP_011538689.1 | 6 | 14097 | |||
XM_017016969.1 | XP_016872458.1 | 15 | 11142 | |||
XM_017016970.1 | XP_016872459.1 | 14 | 10958 | |||
GenBank mRNA | AK025783.1 | - | 1 | 2154 | ||
AK096575.1 | - | 1 | 2746 | |||
AK123570.1 | - | 1 | 3769 | |||
AL833070.2 | - | 1 | 4538 | |||
BX647371.1 | - | 1 | 5147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
