siRNA Details
Gene Symbol: FRMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300832.1 | NP_001287761.1 | 9 | 1114 | ||
NM_001300833.1 | NP_001287762.1 | 9 | 1180 | |||
NM_031904.3 | NP_114110.1 | 10 | 1282 | |||
GenBank mRNA | AF334946.1 | - | 11 | 1216 | ||
AK074850.1 | - | 9 | 1047 | |||
AK291678.1 | - | 10 | 1195 | |||
AK304363.1 | - | 9 | 1093 | |||
BC044658.1 | - | 10 | 1200 | 5759383 | ||
BC051695.1 | - | 10 | 1195 | 5298820 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
