siRNA Details
Gene Symbol: FRMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300832.1 | NP_001287761.1 | 10 | 3514 | ||
NM_001300833.1 | NP_001287762.1 | 10 | 3580 | |||
NM_031904.3 | NP_114110.1 | 11 | 3682 | |||
GenBank mRNA | AK056778.1 | - | 1435 | |||
AK074850.1 | - | 10 | 3448 | |||
BC044658.1 | - | 11 | 3601 | 5759383 | ||
BC051695.1 | - | 11 | 3596 | 5298820 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
