siRNA Details
Gene Symbol: MAPK8IP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040439.1 | NP_001035529.1 | 6, 7 | 1117 | ||
NM_001318852.1 | NP_001305781.1 | 6, 7 | 1157 | |||
NM_015133.4 | NP_055948.2 | 6, 7 | 1154 | |||
XM_005255190.2 | XP_005255247.1 | 6, 7 | 1157 | |||
XM_006720869.2 | XP_006720932.1 | 6, 7 | 2587 | |||
XM_011522429.2 | XP_011520731.1 | 7, 8 | 1181 | |||
XM_011522430.2 | XP_011520732.1 | 7, 8 | 1181 | |||
XM_011522431.2 | XP_011520733.1 | 7, 8 | 1181 | |||
XM_011522432.2 | XP_011520734.1 | 7, 8 | 1181 | |||
XM_011522433.2 | XP_011520735.1 | 2, 3 | 1804 | |||
XM_017023079.1 | XP_016878568.1 | 6, 7 | 1157 | |||
XM_017023080.1 | XP_016878569.1 | 6, 7 | 1157 | |||
XM_017023081.1 | XP_016878570.1 | 5, 6 | 629 | |||
GenBank mRNA | AB028989.2 | - | 6, 7 | 1117 | ||
BC137123.1 | - | 6, 7 | 1069 | 9021120 | ||
BC137124.1 | - | 6, 7 | 1069 | 9021121 | ||
BC144486.1 | - | 6, 7 | 1072 | 9053013 | ||
BC150266.1 | - | 6, 7 | 1117 | 9007222 | ||
CA776361.1 | - | 1, 2 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
