siRNA Details
Gene Symbol: MAPK8IP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040439.1 | NP_001035529.1 | 27 | 3598 | ||
NM_001318852.1 | NP_001305781.1 | 28 | 3656 | |||
NM_015133.4 | NP_055948.2 | 28 | 3653 | |||
XM_005255190.2 | XP_005255247.1 | 27 | 3611 | |||
XM_006720869.2 | XP_006720932.1 | 28 | 5086 | |||
XM_011522429.2 | XP_011520731.1 | 29 | 3680 | |||
XM_011522430.2 | XP_011520732.1 | 28 | 3662 | |||
XM_011522431.2 | XP_011520733.1 | 29 | 3653 | |||
XM_011522432.2 | XP_011520734.1 | 28 | 3635 | |||
XM_011522433.2 | XP_011520735.1 | 24 | 4303 | |||
XM_017023079.1 | XP_016878568.1 | 27 | 3638 | |||
XM_017023080.1 | XP_016878569.1 | 28 | 3629 | |||
XM_017023081.1 | XP_016878570.1 | 27 | 3128 | |||
GenBank mRNA | AB028989.2 | - | 28 | 3616 | ||
BC137123.1 | - | 28 | 3568 | 9021120 | ||
BC137124.1 | - | 28 | 3568 | 9021121 | ||
BC144486.1 | - | 28 | 3571 | 9053013 | ||
BC150266.1 | - | 28 | 3616 | 9007222 | ||
BQ961808.1 | - | 5 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
