siRNA Details
Gene Symbol: HN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002032.2 | NP_001002032.1 | 2 | 471 | ||
NM_001002033.2 | NP_001002033.1 | 3 | 566 | |||
NM_001288609.1 | NP_001275538.1 | 2 | 1155 | |||
NM_001288610.1 | NP_001275539.1 | 2 | 518 | |||
NM_001288611.1 | NP_001275540.1 | 2 | 471 | |||
NM_016185.3 | NP_057269.1 | 2 | 471 | |||
NR_109933.1 | 2 | 471 | ||||
GenBank mRNA | AF086910.2 | - | 2 | 471 | ||
AF177862.1 | - | 2 | 288 | |||
AF266846.1 | - | 2 | 300 | |||
AK223321.1 | - | 2 | 300 | |||
AK308253.1 | - | 2 | 518 | |||
AK312613.1 | - | 2 | 300 | |||
AY322169.1 | - | 2 | 463 | |||
AY322170.1 | - | 3 | 558 | |||
BC001420.2 | - | 2 | 289 | 3140086 | ||
BC039343.1 | - | 2 | 1157 | 5269120 | ||
BG779109.1 | - | 2 | 301 | |||
CD359142.1 | - | 467 | ||||
DA604735.1 | - | 2 | 300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
