siRNA Details
Gene Symbol: HN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002032.2 | NP_001002032.1 | 4 | 961 | ||
NM_001002033.2 | NP_001002033.1 | 6 | 1075 | |||
NM_001288609.1 | NP_001275538.1 | 5 | 1664 | |||
NM_001288610.1 | NP_001275539.1 | 5 | 1027 | |||
NM_001288611.1 | NP_001275540.1 | 5 | 984 | |||
NM_016185.3 | NP_057269.1 | 5 | 980 | |||
NR_109933.1 | 5 | 1016 | ||||
GenBank mRNA | AF060925.1 | - | 1 | 246 | ||
AF086910.2 | - | 5 | 980 | |||
AF266846.1 | - | 5 | 809 | |||
AK223321.1 | - | 5 | 809 | |||
AK308253.1 | - | 5 | 1026 | |||
AY322169.1 | - | 4 | 953 | |||
AY322170.1 | - | 6 | 1067 | |||
BC039343.1 | - | 5 | 1666 | 5269120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
