siRNA Details
Gene Symbol: LINC01152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_110124.1 | 3 | 2650 | |||
NR_110132.1 | 3 | 2140 | ||||
GenBank mRNA | D43770.1 | - | LIT1584 CMPD |
3 | 2488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
