siRNA Details
Gene Symbol: C6orf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008703.2 | NP_001008703.1 | 5 | 856 | ||
NM_001008704.2 | NP_001008704.1 | 5 | 796 | |||
NM_001287396.1 | NP_001274325.1 | 5 | 1048 | |||
NM_001287397.1 | NP_001274326.1 | 4 | 1082 | |||
NM_001287398.1 | NP_001274327.1 | 4 | 830 | |||
NM_178508.4 | NP_848603.2 | 5 | 1108 | |||
GenBank mRNA | AK290164.1 | - | 5 | 790 | ||
BC023627.2 | - | 5 | 728 | 4764944 | ||
BC047919.1 | - | 5 | 989 | 6190218 | ||
BM474526.1 | - | 4 | 748 | |||
BU956518.1 | - | 4 | 595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
