siRNA Details
Gene Symbol: C6orf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008703.2 | NP_001008703.1 | 5 | 657 | ||
NM_001008704.2 | NP_001008704.1 | 5 | 597 | |||
NM_001287396.1 | NP_001274325.1 | 5 | 849 | |||
NM_001287397.1 | NP_001274326.1 | 4 | 883 | |||
NM_001287398.1 | NP_001274327.1 | 4 | 631 | |||
NM_178508.4 | NP_848603.2 | 5 | 909 | |||
GenBank mRNA | AK290164.1 | - | 5 | 591 | ||
BC023627.2 | - | 5 | 529 | 4764944 | ||
BC047919.1 | - | 5 | 790 | 6190218 | ||
BM474526.1 | - | 4 | 548 | |||
BU956518.1 | - | 4 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
