siRNA Details
Gene Symbol: LOC403323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122076.1 | 2 | 598 | |||
NR_122077.1 | 2 | 708 | ||||
GenBank mRNA | AK000451.1 | - | 2 | 708 | ||
AK309896.1 | - | 2 | 218 | |||
DB521113.1 | - | 2 | 249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
