siRNA Details
Gene Symbol: LOC403323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122076.1 | 2 | 622 | |||
NR_122077.1 | 2 | 732 | ||||
GenBank mRNA | AK000451.1 | - | 2 | 732 | ||
AK309896.1 | - | 2 | 242 | |||
DB521113.1 | - | 2 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
