siRNA Details
Gene Symbol: HIST1H2BH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003524.2 | NP_003515.1 | 1 | 91 | ||
GenBank mRNA | AK312052.1 | - | 1 | 128 | ||
BC096116.3 | - | 1 | 91 | 40002316 | ||
BC096117.1 | - | 1 | 91 | 40002317 | ||
BC096118.1 | - | 1 | 91 | 40002318 | ||
BC096119.1 | - | 1 | 91 | 40002319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
