siRNA Details
Gene Symbol: VPS13D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015378.3 | NP_056193.2 | 21 | 5571 | ||
NM_018156.3 | NP_060626.2 | 21 | 5571 | |||
GenBank mRNA | AB007922.3 | - | 3 | 1881 | ||
AJ608774.1 | - | 21 | 5543 | |||
AJ608775.1 | - | 21 | 5543 | |||
AL162331.1 | - | 21 | 5543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
