siRNA Details
Gene Symbol: VPS13D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015378.3 | NP_056193.2 | 32, 33 | 7710 | ||
NM_018156.3 | NP_060626.2 | 32, 33 | 7710 | |||
GenBank mRNA | AB007922.3 | - | 14, 15 | 4020 | ||
AJ608774.1 | - | 32, 33 | 7682 | |||
AJ608775.1 | - | 32, 33 | 7682 | |||
AK001481.1 | - | 5, 6 | 830 | |||
AK125118.1 | - | 8, 9 | 1440 | |||
BX641035.1 | - | 7, 8 | 1274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
