siRNA Details
Gene Symbol: FAM110D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024869.2 | NP_079145.2 | 2 | 441 | ||
GenBank mRNA | AK024112.1 | - | 2 | 441 | ||
AK290715.1 | - | 2 | 381 | |||
AU119858.1 | - | 2 | 441 | |||
BC025658.1 | - | 2 | 427 | 5198480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
