siRNA Details
Gene Symbol: FAM149A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006655.2 | NP_001006656.1 | 12, 13 | 1661 | ||
NM_015398.2 | NP_056213.1 | 13, 14 | 1960 | |||
XM_011531835.2 | XP_011530137.1 | 13, 14 | 1918 | |||
XM_017007997.1 | XP_016863486.1 | 13, 14 | 2503 | |||
XM_017008002.1 | XP_016863491.1 | 13, 14 | 1921 | |||
XM_017008003.1 | XP_016863492.1 | 13, 14 | 1777 | |||
XM_017008004.1 | XP_016863493.1 | 13, 14 | 1829 | |||
XM_017008005.1 | XP_016863494.1 | 13, 14 | 1934 | |||
XM_017008006.1 | XP_016863495.1 | 13, 14 | 1826 | |||
GenBank mRNA | AF173894.1 | - | 12, 13 | 1597 | ||
AK057166.1 | - | 13, 14 | 1790 | |||
AL080065.1 | - | 7, 8 | 1006 | |||
BC110816.1 | - | 13, 14 | 1862 | |||
BC136792.1 | - | 10, 11 | 1406 | 9020782 | ||
BC142992.1 | - | 13, 14 | 1914 | 8860446 | ||
BC144411.1 | - | 10, 11 | 1403 | 9052932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
