siRNA Details
Gene Symbol: RXRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291920.1 | NP_001278849.1 | 10 | 5651 | ||
NM_001291921.1 | NP_001278850.1 | 9 | 5442 | |||
NM_002957.5 | NP_002948.1 | 10 | 4979 | |||
GenBank mRNA | AF052092.1 | - | 1 | 987 | ||
BC007925.1 | - | 1 | 1902 | |||
BC063827.1 | - | 10 | 4968 | |||
U66306.1 | - | 1 | 3230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
